Learn More
Polypropylene Specimen Containers
Product Code. 10072734
Change view
Click to view available options
Capacity (Metric):
125 mL
Unit Size:
Pack of 500
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10072734 125 mL Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10072734 Supplier Copan Italia Supplier No. D103832B
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Closure Color Green
Closure Included Yes
Closure Material Polyethylene
Closure Type Cap
Diameter (Metric) Outside 63 mm
Quantity 500 Pk.
Capacity (Metric) 125 mL
Color Transparent
Product Type Specimen Container
Material Polypropylene
Graduations Yes
Capacity 125 mL