Learn More
Hirschmann™ Polypropylene Stoppers
A simple and economical choice for the closure of vessels.
Brand: Hirschmann™ 8900280
Additional Details : Weight : 0.25000kg
Description
- Manufactured from polypropylene
- With standard ground
- Available in a range of sizes
Specifications
Natural | |
Closing vessels | |
NS 24/29 | |
Polypropylene |