Learn More
Huenersdorf™ Polypropylene Storage Bin
Specifications
Specifications
| Type | Storage Bin |
| Material | Polypropylene |
| Color | Blue |
| Shape | Rectangular |
| Mounting Type | Stackable |
| Capacity (Metric) | 0.73 L, 136 kg |
| For Use With (Application) | Food Safe |
| Quantity | 1/Pk. |
| Dimensions (L x W x H) | 165 x 100 x 75 mm |
| Length (Metric) | 164 mm |
| Show More |