Learn More
Masterflex™ Polypropylene Straight Male Pipe Adapter
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings
Brand: Masterflex™ 30815-25
Additional Details : Weight : 0.69000kg
Description
- Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal
- PVDF Composition
Specifications
Straight Male Pipe Adapter | |
Not Autoclavable | |
1 11/32 in. | |
3/8 in. | |
Ethylene Oxide | |
13/16 in. |
Compression x NPT(M) Connection | |
Polypropylene | |
3/8 in. | |
3/8 in. | |
1/4 in. | |
White |