Learn More
Masterflex™ Tee-Connector Compression Fitting, polypropylene
Product Code. 11751759
Click to view available options
For Use With:
1/2 in. tubing O.D.
1/4 in. tubing O.D.
1/8 in. tubing O.D.
3/8 in. tubing O.D.
Unit Size:
Pack of 10
Product Code. 11751759
Brand: Masterflex™ 0638205
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Connect rigid tubing systems with these tee connectors. Polypropylene Tee-Connector Compression Fittings form a tight seal and withstand higher pressures than barbed fittings.
Assemble fittings to tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
  • Excellent chemical resistance
  • Rated to 220 psi maximum at 25 °C
  • Usable from 0 to 100 °C
  • Sterilization: Ethylene Oxide (ETO)
Specifications
Material Polypropylene
For Use With 1/8 in. tubing O.D.
Min. Operating Temperature (Metric) 0°C
Packaging Quantity 10 Pack
Port Size 3/32 in. orifice; 1-1/8 x 3/4 in.
Maximum Pressure 220 psi
Max. Operating Temperature (Metric) 100°C
Product Type Compression Fitting