Learn More
Kartell™ Polypropylene 'T' Shaped Untapered Connectors
The ideal leak-proof solution for connecting tubes. Kartell™ Polypropylene 'T' Shaped Untapered Connectors are useful for connecting tubes of various diameters.
Brand:  Kartell™ 7052100
Product Code. 12349528
  • € 16.40 / Pack of 10
Only null left
Estimated Shipment: 01-08-2025
Description
  • Excellent chemical and temperature resistance
  • With serrations to hold hoses tightly and prevent slipping
  • Autoclavable
  • Untapered