Learn More
Kartell™ Polypropylene 'T' Shaped Untapered Connectors
The ideal leak-proof solution for connecting tubes. Kartell™ Polypropylene 'T' Shaped Untapered Connectors are useful for connecting tubes of various diameters.
Brand:  Kartell™ 7052100
Product Code. 12349528
  • ONLINE EXCLUSIVE PRICE
    € 12.55
    € 11.88 / Pack of 10
    Save € 0.67
    5% Off
Only null left
Estimated Shipment: 28-01-2026
Add to basket
Description
Description
  • Excellent chemical and temperature resistance
  • With serrations to hold hoses tightly and prevent slipping
  • Autoclavable
  • Untapered