Learn More
Buerkle™ Polypropylene Thread Adapter
Buerkle™ Polypropylene Thread Adapter facilitates the secure connection of pumps and discharge cocks to a variety of containers.
Brand:  Buerkle™ 0600-2002
21.98 EUR valid until 2024-12-31
Use promo code "23267" to get your promotional price.
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10483472
  • € 23.95 / Each
In Stock
to see stock.
Add to basket
Description
Description
  • Constructed of polypropylene
  • Interchange the connections to different adapters
Specifications
Specifications
Thread Adapter
Polypropylene
Internal: 71 mm - 2 in. (BSP)
Brown