Learn More
Buerkle™ Threaded Cap with Outer Thread
Product Code. 10599032 Shop All Bürkle Products
Click to view available options
Thread Size:
1 in.
1/2 in.
3/4 in.
Unit Size:
Each
Product Code. 10599032
Brand: Buerkle™ 85360012
Only null left
Add to basket
Add to basket
A stopper manufactured from polypropylene.
  • Polypropylene construction
  • Autoclavable to 121°C
  • Safe for use with foodstuffs
  • Supplied in packs of 10
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EU Foodstuffs
Autoclavable Autoclavable
Material Polypropylene
Thread Size 1/2 in.
Product Type Closing Stopper Outer Thread