missing translation for 'onlineSavingsMsg'
Learn More

VITLAB™ PVC Tray

Store, carry and organize a variety of lab and office supplies. VITLAB™ PVC Trays are constructed of white PVC for a durable and economical storage option. These molded plastic dishes have rounded edges and are easy to clean. Available in several sizes.

Brand:  VITLAB™ 80285

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 10514261

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

These molded one-piece trays are made of rigid plastic and are well suited for general purpose storage, transport and organization. They are an economical, all-purpose option for labs and work areas.
  • Constructed of rigid polyvinyl chloride (PVC)
    • Strong but lightweight plastic
    • Biologically and chemically resistant (e.g. acids, salts, bases, fats, and alcohols)
  • Rounded corners and edges for comfortable handling
  • One-piece, molded dishes with no seams is easy to clean
Specifications

Specifications

520 x 420 mm
100 mm
PVC
420 mm
Rectangular
White
520 mm
Dish
1/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers