Learn More
Greiner Bio-One Polypropylene Tubes, Round Bottom, Sterile, with White Screw Cap
Description
- Durable polypropylene tubes.
- Sterile.
- Include screw cap closures.
Specifications
Specifications
| Closure Color | White |
| Capacity (Metric) | 12 mL |
| Length (Metric) | 100 mm |
| Material | Polypropylene |
| Quantity | 900/Pk. |
| Sterility | Sterile |
| Diameter (Metric) | 16.8 mm |
| Closure Type | Screw Cap |