missing translation for 'onlineSavingsMsg'
Learn More

Ark Plas Products™ Polypropylene Tubing

Designed for indoor/outdoor applications requiring more rigid tubing. Ark Plas Products™ Polypropylene Tubing is extruded using state of the art in-line process monitoring equipment to ensure the highest quality.

Brand:  Ark Plas Products™ TFX78-TP0

Additional Details : Weight : 0.96300kg

 View more versions of this product

Product Code. 11733904

  • € 103.16 / Each


In stock, ready to ship: 3
Add to basket

Description

Description

Ark Plas tubing is available in a wide range of chemical and physical characteristics to fit most any application, dependably and economically.
  • Standard tubing is shipped bulk on cardboard spools
  • Manufactured in almost every increment from 1/16 to 3/8 in. I.D.
  • Metric sized tubing is also available from 2.5 to 6.5mm I.D.
  • Colors include Natural, Opaque Black, Blue, Red, Green, Yellow and White; and Transparent Blue, Red, Green and Yellow
Specifications

Specifications

Natural
6.4 mm
9.6 mm
1.65 mm
Autoclavable
Polypropylene
500 m
1 Pack
Videos
SDS
Documents