missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ Polypropylene Water Filtration Elements

Brand:  Masterflex™ 29650-54

Additional Details : Weight : 0.38000kg

 View more versions of this product

Product Code. 11754603

Quantity Price
1 € 146.00 / Each
10 € 127.94 / Each
Estimated Shipment
16-02-2021
Add to basket


Description

Description

Specifications

Specifications

52°C
Water Filtration Elements
25.4mm
Polypropylene
Water Filtration Elements
50μm
Videos
SDS
Documents