missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ Polypropylene Water Filtration Elements

Brand:  Masterflex™ 29650-50

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11764603

Quantity Price
1 € 147.00 / Each
10 € 128.82 / Each
Estimated Shipment
11-03-2021
Add to basket


Description

Description

Specifications

Specifications

52°C
Water Filtration Elements
25.4mm
Polypropylene
Water Filtration Elements
100μm
Videos
SDS
Documents