missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyquaternium-37, TRC
Specifications
Specifications
| Chemical Name or Material | Polyquaternium-37 |
| CAS | 26161-33-1 |
| Molecular Formula | (C9H18ClNO2)x |
| Quantity | 1 g |
| Synonym | Choline Chloride Methacrylate PolymersN,N,N-Trimethyl-2-[(2-methyl-1-oxo-2-propenyl)oxy]ethanaminium Chloride HomopolymerMethacrylic Acid Ester With Choline Chloride Polymers, (Methacryloxyethyl)trimethylammonium Chloride Polymer, 2-(Methacryloyloxy)ethyl Trimethylammonium Chloride Homopolymer, 2-(Trimethylammonium)ethyl Methacrylate Chloride Homopolymer, Alcostat 567, Cosmedia CTH(E), Dimethylaminoethyl Methacrylate Methyl Chloride Homopolymer, Dimethylaminoethyl Methacrylate-methyl Chloride Copolymer, Evagrowth C 104G, Flocogil C 4, Himoloc MP 173H, Himoloc NR 70, Himoloc NR 703, K 415, Kayafloc C 599, Light Ester DQ 100 Homopolymer, Methacroylcholine Chloride Homopolymer, PMETAC, Poly(2-methacryloxyethyltrimethylammonium Chloride), Poly(2-methacryloyloxyethyltrimethylammonium Chloride), Poly(2-trimethylammonioethyl Methacrylate Chloride), Poly(N,N,N-trimethylaminoethyl Methacrylate Chloride), Poly(dimethylaminoethyl Methacrylate Methyl Chloride), Poly(methacryloxyethyltrimethylammonium Chloride), Poly(methacryloyloxyethyltrimethylammonium Chloride), Poly(methylacryloyloxyethyltrimethyl Sodium Chloride), Poly(β-methacryloxyethyltrimethylammonium Chloride), PolyTMMC, Poly[(trimethylammonio)ethyl Methacrylate Chloride], Poly[N,N,N-trimethyl-N-(2-methacryloxyethyl)ammonium Chloride], Poly[[β-(methacryloyloxy)ethyl]trimethylammonium Chloride], Poly[trimethyl(β-methacryloyloxyethyl)ammonium Chloride], Poly[trimethyl[2-(methacryloyloxy)ethyl]ammonium Chloride], Praestol 444K, QDM WFixer 013, Rheocare CTH(E), Rheocare Ultragel 300, Salcare SC 95, Sanfloc C 009P, Shallol, Synthalen CR, Takiflock C 409, Trimethylaminoethyl Methacrylate Chloride Homopolymer, Trimethylammonioethyl Methacrylate Chloride Homopolymer, Trimethylammonioethyl Methacrylate Chloride Polymer, Ultragel 300, Zetag 88N, [2-(Methacroyloxy)ethyl]trimethylammonium Chloride Homopolymer, β-Methacryloyloxyethyltrimethylammonium Chloride Polymer |
| InChI Formula | InChI=1S/C9H18NO2.ClH/c1-8(2)9(11)12-7-6-10(3,4)5;/h1,6-7H2,2-5H3;1H/q+1;/p-1 |
| SMILES | O=C(C(C)=C)OCC[N+](C)(C)C.[Cl-] |
| IUPAC Name | 2-(methacryloyloxy)-N,N,N-trimethylethan-1-aminium chloride |
| Molecular Weight (g/mol) | 207.7 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?