Learn More
Kartell™ Polystyrene Biological Specimen Containers
Product Code. 10355631
Click to view available options
Capacity (Metric):
60 mL
200 mL
Diameter (Metric):
39 mm
66 mm
Height (Metric):
65 mm
86 mm
Unit Size:
Pack of 200
Pack of 400
Product Code. 10355631
Brand: Kartell™ 0563100
Manufactured with crystal clear polystyrene for optimal visibility. Kartell™ Polystyrene Biological Specimen Containers are available with polyethylene and polypropylene color coded screw caps.
  • Individually wrapped
  • CE marked;IVD approved
Specifications
Product Type Biological Specimen Container
Closure Included Yes
Material Polystyrene
Sterility Aseptic
Capacity (Metric) 60 mL
Certifications/Compliance CE and IVD
Closure Color Red
Closure Material Polypropylene
Closure Type Screw Cap
Diameter (Metric) 39 mm
For Use With (Application) Urine and biological specimen collection
Height (Metric) 65 mm
Includes Polypropylene Screw Cap
Label Yes
No. per Pack 400/Pk.