missing translation for 'onlineSavingsMsg'
Learn More

VITLAB™ Polystyrene Lid

Protect the contents of instrument trays from dust and contamination. VITLAB™ Polystyrene Lid is constructed of crystal clear polystyrene to keep contents visible at all times. Features a knob handle, rounded edges and a smooth, easy-to-clean surface. Available in several sizes.

Brand:  VITLAB™ 79990

Additional Details : Weight : 1.67000kg

 View more versions of this product

Product Code. 15752006

  • € 60.68 / Pack of 5


Estimated Shipment
30-11-2020
Add to basket

Description

Description

  • Constructed of rigid polystyrene for a strong, durable lid
  • Rounded corners and a knob handle for comfortable handling
  • Completely transparent lid for easy identification of contents
  • Protects contents from dust and other contaminants
  • Smooth surface is easy to clean
Specifications

Specifications

Instrument Tray Lid
Tray number 72498
5/Pk.
340 x 245 mm
340 mm
Instrument Tray Lid
340 x 245 mm
Handle
Rectangular
Clear
Polystyrene
245 mm
Videos
SDS
Documents