Learn More
Multiroir™ Polystyrene Rectangular Trays
Click to view available options
Height (Metric):
34 mm (Inner), 37 mm (Outer)
40 mm (Inner), 42 mm (Outer)
42 mm
Color:
Blue
Green
Red
White
Yellow
Unit Size:
Each
Description
- Multi-purpose
- Polystyrene
- Rectangular
Specifications
Specifications
| Color | Yellow |
| Height (Metric) | 34 mm (Inner), 37 mm (Outer) |
| Length (Metric) | 175 mm (Inner), 205 mm (Outer) |
| Material | Polystyrene |
| Type | Tray |
| Width (Metric) | 100 mm (Inner), 130 mm (Outer) |
| No. per Pack | 1 Pack |
| Shape | Rectangular |