missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Safety Products™ Polytril™ Air 3/4 Gloves

Catalog Number 12695433
Click to view available options
Size:
7
8
This item is not returnable. View return policy
This item is not returnable. View return policy

Resistant polyamide knitted gloves with nitrile foam-coated palm and fingers provide good grip for precision work, especially in greasy, damp, or dirty environments. Honeywell Safety Products™ Polytril™ Air 3/4 Gloves are breathable, comfortable and provide excellent resistance.

ul>
  • Black, non-pilling polyamide knitted glove suitable for precision work.
  • Suitable for handling fine products or dirty objects in greasy, damp or dirty environments.
  • Excellent abrasion and tear resistance.
  • Black nitrile foam coating on palm and fingers provides oil impermeability, abrasion resistance, and good grip.
  • Tight knit provides comfort, support, dexterity, touch and limits dirt penetration.
  • Easily laundered for extended use.
  • Ventilated back optimizes breathability.
  • Elastic wrist.
  • Black color suitable for work in soiled environments.
  • TRUSTED_SUSTAINABILITY
    Certifications/Compliance CE CATEGORY II Intermediary risks, EN 420-2003 Basic standards, EN 388-2003 Mechanical hazard, EC Category PPE 2, Quality Assurance ISO 9001 / 2000, EC Certificate Number N°0075/797/162/05/07/0193, EC Attestation Number N°0075/797/162/05/07/0193
    Classification Dexterity: 5
    Coated Area Palm, Fingers
    Construction Seamless Knit
    Description Gauge 13
    Coating Material Nitrile Foam
    Color Black
    Cut Resistance Rating 1
    Material Nitrile, Polyamide
    Product Type Gloves
    Size 7
    For Use With (Application) Automotive, Aviation, Logistics, Maintenance and Transportation industries.
    Puncture Rating 1
    Quantity 10Pair/PK
    Tear Rating 2
    Length (Metric) 24.5 cm
    Show More Show Less
    missing translation for 'provideContentCorrection'

    missing translation for 'productTitle'