Learn More
Kautex™ PVC Wide Neck Square Container, Transparent, Without Closure
Fill and clean these wide neck PVC containers with ease due to the roomy neck configuration. Supplied without closure.
Brand: Kautex™ 2000070596
Additional Details : Weight : 0.01000kg
Description
- Member of Series 310 container family
- Closure available separately
Specifications
Wide Neck Container | |
Closure, Foam Insert | |
Transparent | |
Packaging, Dispensing, Sending Samples, Storage, Mailing | |
480 | |
No |
71 mm | |
100 mL | |
31 mm | |
PVC | |
480/Cs. | |
Square |