Learn More
Kautex™ PVC Wide Neck Square Container, Transparent, Without Closure
Product Code. 15961451
Click to view available options
Capacity (Metric):
100 mL
200 mL
300 mL
500 mL
750 mL
1000 mL
2000 mL
Height (Metric):
71 mm
83.5 mm
87 mm
94 mm
98.5 mm
105.5 mm
108 mm
118.5 mm
120 mm
143 mm
187 mm
Unit Size:
Each
Pack of 114
Pack of 126
Pack of 162
Pack of 21
Pack of 288
Pack of 44
Pack of 48
Pack of 480
Pack of 80
Pack of 96
Product Code. 15961451
Brand: Kautex™ 2000084419
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Fill and clean these wide neck PVC containers with ease due to the roomy neck configuration. Supplied without closure.
  • Member of Series 310 container family
  • Closure available separately
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 100 mL
Color Transparent
Diameter (Metric) Inner Neck 31 mm
For Use With (Application) Packaging, Dispensing, Sending Samples, Storage, Mailing
Material PVC
No. per Case 480
Quantity 480/Cs.
Closure Included No
Shape Square
Height (Metric) 83.5 mm
Product Type Wide Neck Container
For Use With (Equipment) Closure