Learn More
Kautex™ PVC Wide Neck Square Container, Transparent, Without Closure
Click to view available options
Capacity (Metric):
100 mL
200 mL
300 mL
500 mL
750 mL
1000 mL
2000 mL
Height (Metric):
71 mm
83.5 mm
87 mm
94 mm
98.5 mm
105.5 mm
108 mm
118.5 mm
120 mm
143 mm
187 mm
Unit Size:
Each
Pack of 114
Pack of 126
Pack of 162
Pack of 21
Pack of 288
Pack of 44
Pack of 48
Pack of 480
Pack of 80
Pack of 96
Description
- Member of Series 310 container family
- Closure available separately
Specifications
Specifications
| Capacity (Metric) | 750 mL |
| Color | Transparent |
| Diameter (Metric) Inner Neck | 69 mm |
| For Use With (Application) | Packaging, Dispensing, Sending Samples, Storage, Mailing |
| Material | PVC |
| No. per Case | 114 |
| Quantity | 114/Cs. |
| Closure Included | No |
| Shape | Square |
| Height (Metric) | 120 mm |
| Show More |