missing translation for 'onlineSavingsMsg'
Learn More

Polyvinylpyrrolidone, M.W. 8,000, Thermo Scientific Chemicals

Catalog Number p-7051598
40.20 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
9003-39-8
(C6H9NO)n
111.14
MFCD01076626
WHNWPMSKXPGLAX-UHFFFAOYSA-N
n-vinyl-2-pyrrolidone, n-vinylpyrrolidone, 1-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, n-vinyl-2-pyrrolidinone, vinylpyrrolidone, 2-pyrrolidinone, 1-ethenyl, 1-vinyl-2-pyrrolidinone, n-vinylpyrrolidinone, 1-vinylpyrrolidone
6917
CHEBI:82551
1-ethenylpyrrolidin-2-one
*-CC(-*)N1CCCC1=O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
9003-39-8
(C6H9NO)n
111.14
MFCD01076626
WHNWPMSKXPGLAX-UHFFFAOYSA-N
n-vinyl-2-pyrrolidone, n-vinylpyrrolidone, 1-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, n-vinyl-2-pyrrolidinone, vinylpyrrolidone, 2-pyrrolidinone, 1-ethenyl, 1-vinyl-2-pyrrolidinone, n-vinylpyrrolidinone, 1-vinylpyrrolidone
6917
CHEBI:82551
1-ethenylpyrrolidin-2-one
*-CC(-*)N1CCCC1=O

A cryoprotectant, also useful in plant cell culture research

A cryoprotectant, also useful in plant cell culture research Polyvinylpyrrolidone is used as a cryoprotectant. It is used as a binder for tablets in pharmaceuticals. It is added to iodine and forms a complex povidone-iodine, which is having disinfectant properties, used in various products like solutions, ointment and pessaries. It is used as an adhesive in glue stick, emulsifier, additive for batteries, ceramics, fiberglass, inks, and inkjet paper, thickening agent in tooth whitening gels and to increase resolution in photoresists for cathode ray tubes. Further, it is used in personal care products such as shampoos, toothpastes, hair sprays and hair gels. In addition to this, it finds application in contact lens solutions, disintegrant for solution polymerization and in steel-quenching solutions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Powder
Melting Point 130°C
Merck Index 14,7697
Solubility Information Soluble in water,alcohol,chlorinated hydrocarbons,amines,nitro paraffins,lower wt fatty acids and chloroform. Insoluble in ether.
Quantity 100 g
Odor Odorless
Chemical Name or Material Polyvinylpyrrolidone, M.W. 8,000

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'