Learn More
MedChemExpress Pomalidomide-PEG4-Ph-NH2
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Specifications
Specifications
| Molecular Weight (g/mol) | 540.57 g/mol |
| CAS | 1818885-63-0 |
| Content And Storage | Powder -20°C 3 years, 4°C 2 years ∣ In solvent -80°C 6 months, -20°C 1 month |
| Shipping Condition | Room temperature |
| Quantity | 100 mg |
| Synonym | Cereblon Ligand-Linker Conjugates 9 ∣ E3 Ligase Ligand-Linker Conjugates 2 |
| Solubility | DMSO : ≥ 50 mg/mL (92.49 mM) ∣ H2O : < 0.1 mg/mL (insoluble) |
| Target | HBV |