missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POMC Mouse anti-Human, Clone: CLIP/1407, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB14873.100ug
This item is not returnable.
View return policy
Description
Mouse monoclonal antibody raised against synthetic peptide of human POMC.
- High specificity
- High reactivity
Specifications
| POMC | |
| Monoclonal | |
| Unconjugated | |
| Flow Cytometry (0.5-1 ug/106 cells) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| P01189 | |
| POMC | |
| A synthetic peptide corresponding to amino acids 25-39 of human POMC. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| CLIP/1407 | |
| Mouse monoclonal antibody raised against synthetic peptide of human POMC. | |
| In 10 mM PBS (0.05% BSA, 0.05% sodium azide). | |
| ACTH/CLIP/LPH/MSH/NPP/POC | |
| Mouse | |
| Protein A/G purification | |
| RUO | |
| 5443 | |
| Store at 4°C. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction