missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ POMK Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA543488
This item is not returnable.
View return policy
Description
Peptide sequence: CEELRTEVRQ LKRVGEGAVK RVFLSEWKEH KVALSQLTSL EMKDDFLHGL Sequence homology: Cow: 93%; Dog: 86%; Guinea Pig: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 93%.
This gene encodes a protein that may be involved in the presentation of the laminin-binding O-linked carbohydrate chain of alpha-dystroglycan (a-DG), which forms transmembrane linkages between the extracellular matrix and the exoskeleton. Some pathogens use this O-linked carbohydrate unit for host entry. Loss of function compound heterozygous mutations in this gene were found in a human patient affected by the Walker-Warburg syndrome (WWS) phenotype. Mice lacking this gene contain misplaced neurons (heterotopia) in some regions of the brain, possibly from defects in neuronal migration. Alternative splicing of this gene results in multiple transcript variants.
Specifications
| POMK | |
| Polyclonal | |
| Unconjugated | |
| POMK | |
| 4930444A02Rik; MDDGA12; MDDGC12; Pomk; probable inactive protein kinase-like protein SgK196; protein kinase-like protein SgK196; protein kinase-like protein SgK196-like; protein O-mannose kinase; protein-O-mannose kinase; RGD1310810; SGK196; sugen kinase 196 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 74653, 84197 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q3TUA9, Q9H5K3 | |
| POMK | |
| Synthetic peptide directed towards the N-terminal of human FLJ23356 (aa 74-123). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction