missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POMK/SGK196 Antibody (OTI2B10) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72275
This item is not returnable.
View return policy
Description
POMK/SGK196 Monoclonal specifically detects POMK/SGK196 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| POMK/SGK196 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:2000, Immunohistochemistry 1:150 | |
| FLJ23356, MGC126597, protein kinase-like protein SgK196, Sugen kinase 196 | |
| Mouse | |
| 39.9 kDa | |
| 100 μg | |
| Primary | |
| Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI2B10 | |
| Unconjugated | |
| Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
| POMK | |
| Human recombinant protein fragment corresponding to amino acids 141-350 of human SGK196 (NP_115613) produced in E.coli. | |
| Affinity Purified | |
| Protein Kinase | |
| 84197 | |
| Human | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction