Learn More
PON1 Mouse anti-Human, Clone: OTI2A9, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF502612
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The paraoxonase (PON) gene-family in humans has three members, PON1, PON2, and PON3, aligned next to each other on the long arm of chromosome 7q21.3-22.1. PON1 of these is synthesized primarily in the liver and a portion is secreted into the plasma, where it is associated with high density lipoproteins (HDL). And It hydrolyzes the active metabolites of several other organophosphorus insecticides (e.g., chlorpyrifos oxon, diazoxon), as well as nerve agents such as sarin, soman and VX (lethal nerve agent). The PON1 polymorphism may be responsible for neurodegeneration and considered to be an independent risk factor for Parkinson′s disease.Specifications
| PON1 | |
| Monoclonal | |
| Unconjugated | |
| PON1 | |
| ESA, MVCD5, PON | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 5444 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Western Blot | |
| OTI2A9 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P27169 | |
| PON1 | |
| Full length human recombit protein of human PON1 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.