Learn More
Invitrogen™ PON1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120197
Description
Positive test controls include: HepG2, NCI-H460, HT-1080, THP-1, Mouse kidney, Rat kidney. The target is usually found in the following locations: Secreted, extracellular space. Immunogen sequence: LLGMGLALFR NHQSSYQTRL NALREVQPVE LPNCNLVKGI ETGSEDLEIL PNGLAFISSG LKYPGIKSFN PNSPGKILLM DLNEEDPTVL ELGITGSKFD V.
The paraoxonase (PON) gene-family in humans has three members, PON1, PON2, and PON3, aligned next to each other on the long arm of chromosome 7q21.3-22.1. PON1 of these is synthesized primarily in the liver and a portion is secreted into the plasma, where it is associated with high density lipoproteins (HDL). And It hydrolyzes the active metabolites of several other organophosphorus insecticides (e.g., chlorpyrifos oxon, diazoxon), as well as nerve agents such as sarin, soman and VX (lethal nerve agent). The PON1 polymorphism may be responsible for neurodegeneration and considered to be an independent risk factor for Parkinson's disease.
Specifications
| PON1 | |
| Polyclonal | |
| Unconjugated | |
| PON1 | |
| 2810004E20; 6330405I24Rik; A esterase 1; A-esterase 1; A-esterase 2; AI481612; AI786302; Aromatic esterase 1; aromatic esterase 2; arylesterase 1; arylesterase 2; arylesterase 3; arylesterase B-type; EC 3.1.1.2; EC 3.1.1.81; EC 3.1.8.1; ESA; ESAesterase A; esterase A; K-45; MVCD5; paraoxanase-3; paraoxonase 1; paraoxonase 2; paraoxonase 3; paraoxonase B-type; paraoxonase nirs; PON; PON 1; PON 2; Pon1; PON2; PON3; serum aryldiakylphosphatase; serum aryldialkylphosphatase 1; Serum aryldialkylphosphatase 2; serum paraoxonase/arylesterase 1; Serum paraoxonase/arylesterase 2; serum paraoxonase/lactonase 3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 18979, 5444, 84024 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 3.95 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P27169, P52430, P55159 | |
| PON1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 9-109 of human PON1 (NP_000437.3). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.