missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ponceau S, pure

Product Code. p-2372040 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
This item is not returnable. View return policy

Product Code. 10454915

Brand: Thermo Scientific Chemicals 161470100

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6226-79-5 | C22H12N4Na4O13S4 | 760.552 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6226-79-5
Molecular Formula C22H12N4Na4O13S4
Molecular Weight (g/mol) 760.552
MDL Number MFCD00003892
InChI Key VSXKEUCERCWGKF-STNZDNLRSA-J
Synonym C.I. 27195, 3-Hydroxy-4-[2-sulfo-4-(4-sulfophenylazo)phenylazo]-2, 7-naphthalenedisulfonic acid, tetrasodium salt
PubChem CID 11320219
IUPAC Name tetrasodium;(4Z)-3-oxo-4-[[2-sulfonato-4-[(4-sulfonatophenyl)diazenyl]phenyl]hydrazinylidene]naphthalene-2,7-disulfonate
SMILES C1=CC(=CC=C1N=NC2=CC(=C(C=C2)NN=C3C4=C(C=C(C=C4)S(=O)(=O)[O-])C=C(C3=O)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Na+]

Specifications

Dye Content 70% min.
Solubility Information Solubility in water: 10g/L. Other solubilities: insoluble in ether
Infrared Spectrum Authentic
Formula Weight 760.54
Grade Pure
Concentration or Composition (by Analyte or Components) Solubility    •To pass testE1%1cm    •≥350L g−1cm−1Lambda Max. (λ)    •514nm±2nm
Packaging Glass bottle
UV Spectrometry Complies
Color Bordeaux to Brown/Red
Quantity 10 g
Physical Form Crystalline Powder
Chemical Name or Material Ponceau S
Show More Show Less
compliance-icons
Product Identifier
  • Ponceau S
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.