Learn More
Invitrogen™ POPDC3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113905
Description
Immunogen sequence: PEWDSLRPTE EGIFQVTLTA DTDCRYVSWR RKKLYLLFAQ HRYISRLFSV For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 86%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.
POPDC3 is a member of the POP family of proteins containing three putative transmembrane domains. The protein is expressed in cardiac and skeletal muscle and may play an important role in these tissues during development. This gene encodes a member of the POP family of proteins containing three putative transmembrane domains. This gene is expressed in cardiac and skeletal muscle and may play an important role in these tissues during development.
Specifications
| POPDC3 | |
| Polyclonal | |
| Unconjugated | |
| POPDC3 | |
| AA682104; bA355M14.1; MGC22671; POP3; Popdc3; popeye 3; popeye domain containing 3; popeye domain-containing 3; popeye domain-containing protein 3; Popeye protein 3; RP11-99L11.2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 78977 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9ES81 | |
| POPDC3 | |
| Immunogen sequence: PEWDSLRPTE EGIFQVTLTA DTDCRYVSWR RKKLYLLFAQ HRYISRLFSV (aa 184-233). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.