missing translation for 'onlineSavingsMsg'
Learn More

POPE, TRC

Product Code. 30425376
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30425376 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30425376 Supplier Toronto Research Chemicals Supplier No. TRCP68951050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material POPE
CAS 26662-94-2
Molecular Formula C39H76NO8P
Quantity 50 mg
Synonym 1-Palmitoyl-2-oleoyl-sn-glyceryl-3-phosphorylethanolamine, L-alpha-1-Palmitoyl-2-oleoylglycerophosphoethanolamine, 2-Oleoyl-1-palmitoyl-sn-glycero-3-phosphatidylethanolamine
InChI Formula InChI=1S/C39H76NO8P/c1-3-5-7-9-11-13-15-17-18-20-22-24-26-28-30-32-39(42)48-37(36-47-49(43,44)46-34-33-40)35-45-38(41)31-29-27-25-23-21-19-16-14-12-10-8-6-4-2/h17-18,37H,3-16,19-36,40H2,1-2H3,(H,43,44)/b18-17-/t37-/m1/s1
SMILES CCCCCCCCCCCCCCCC(=O)OC[C@H](COP(=O)([O-])OCC[NH3+])OC(=O)CCCCCCC\C=C/CCCCCCCC
IUPAC Name 2-azaniumylethyl [(2R)-3-hexadecanoyloxy-2-[(Z)-octadec-9-enoyl]oxypropyl] phosphate
Molecular Weight (g/mol) 718
Formula Weight 717.5309
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.