Learn More
POR Mouse anti-Human, Clone: OTI3H10, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF500572
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Cytochrome P450 Reductase encodes an endoplasmic reticulum membrane oxidoreductase with an FAD-binding domain and a flavodoxin-like domain. The protein binds two cofactors, FAD and FMN, which allow it to donate electrons directly from NADPH to all microsomal P450 enzymes. Mutations in this gene have been associated with various diseases, including apparent combined P450C17 and P450C21 deficiency, amenorrhea and disordered steroidogenesis, congenital adrenal hyperplasia and Antley-Bixler syndrome.Specifications
| POR | |
| Monoclonal | |
| Unconjugated | |
| POR | |
| CPR, CYPOR, P450R | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 5447 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI3H10 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P16435 | |
| POR | |
| Full-length protein expressed in 293T cell transfected with human POR expression vector | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.