Learn More
Haldenwanger™ Porcelain Buchner Funnel
Click to view available options
Capacity (Metric):
10 mL
10,600 mL
1250 mL
25 mL
3 mL
4300 mL
5800 mL
Diameter (Metric) Plate:
19 mm
29 mm
42 mm
155 mm
250 mm
279 mm
330 mm
Unit Size:
Each
Description
- Available in a variety of sizes
- Available with perforated fixed filter plate
- Complies with DIN VDE 0335, type C 100
Specifications
Specifications
| Capacity (Metric) | 4300 mL |
| Material | Porcelain |
| Height (Metric) | 330 mm |
| Diameter (Metric) Plate | 250 mm |
| Diameter (Metric) Disc | 250 mm |
| Quantity | 1Each |
| Product Type | Buchner Funnel |