Learn More
Haldenwanger™ Porcelain Casserole Basin
Brand:  Haldenwanger™ 17/4
Additional Details : Weight : 0.70000kg
Product Code. 10291001
  • € 255.46 / Each
Estimated Shipment: 18-06-2024
to see stock.
Add to basket
Description
Description
Order Info
CapacityMetric
Specifications
Specifications
1300 mL
Round
90 mm
Porcelain
Evaporating Basin
160 mm