Learn More
Hecht Karl™ Porcelain Mortar
Click to view available options
Diameter (Metric) Outer:
20mm
70mm
130mm
150mm
Unit Size:
Each
Specifications
Specifications
| Capacity (Metric) | 400 mL |
| Diameter (Metric) Outer | 150mm |
| Product Type | Mortar |
| Material | Porcelain |
| Height (Metric) | 65mm |
| Type | Mortar |