missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Porcine IL-4 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC6543U-100UG
This item is not returnable.
View return policy
Description
IL-4 Monoclonal specifically detects IL-4 in Porcine samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| IL-4 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q04745 | |
| IL4 | |
| E. coli-derived recombinant porcine IL-4 His25-Cys133 Accession # Q04745 | |
| RUO | |
| 3565 | |
| Pig | |
| IgG1 |
| Flow Cytometry | |
| 99605 | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| B cell growth factor 1, BCDF, B-cell stimulatory factor 1, BCGF1, BCGF-1, binetrakin, BSF1, BSF-1, IL4, IL-4B_cell stimulatory factor 1, IL4E12, interleukin 4, interleukin-4, Lymphocyte stimulatory factor 1, MGC79402, Pitrakinra | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects porcine IL-4 in direct ELISAs and Western blots. Does not cross-react with recombinant human IL-13 or with recombinant IL-4 from human, canine, bovine, mouse, rat, cotton rat, or feline systems. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction