missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Porcine IL-5 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF3137-SP
This item is not returnable.
View return policy
Description
IL-5 Polyclonal specifically detects IL-5 in Porcine samples. It is validated for Western Blot, Neutralization, Immunocytochemistry.
Specifications
| IL-5 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BCDF mu, B-cell differentiation factor I, BCGFII, EDF, Eo-CSF, Eosinophil differentiation factor, IL5, IL-5T-cell replacing factor, interleukin 5 (colony-stimulating factor, eosinophil), interleukin-5, TRF, TRFB cell differentiation factor I | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3567 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Neutralization, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Neutralization 2-8 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| Q9MYM5 | |
| IL5 | |
| Mouse myeloma cell line NS0-derived recombinant porcine IL-5 Ile20-Ser134 Accession # Q9MYM5 | |
| 25 μg | |
| Primary | |
| Detects porcine IL-5 in direct ELISAs and Western blots. In direct ELISAs, less than 30% cross-reactivity with recombinant canine IL-5 and less than 10% cross-reactivity with recombinant human IL-5, recombinant mouse IL-5, and recombinant rat IL-5 is observed. | |
| Pig | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction