missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Porcine IL-6 Alexa Fluor 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC6861T-100UG
This item is not returnable.
View return policy
Description
IL-6 Monoclonal specifically detects IL-6 in Porcine samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| IL-6 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| P26893 | |
| IL6 | |
| E. coli-derived recombinant porcine IL-6 Pro29-Met212 Accession # P26893 | |
| RUO | |
| 3569 | |
| Pig | |
| IgG2b |
| Flow Cytometry | |
| 90613 | |
| Alexa Fluor 594 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL6, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6, MGI-2A | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects porcine IL-6 in direct ELISAs and Western blots. In direct ELISAs and Western blots, partial cross-reactivity with recombinant human, mouse, canine, equine, and feline IL-6 is observed, and no cross-reactivity with recombinant human (rh) CLC, rhIL-11, rhCNTF, rhCT-1, rhLIF, or rhOSM is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction