missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Porcine TNF-alpha Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 4 publications
Brand: R&D Systems AF690-SP
This item is not returnable.
View return policy
Description
TNF-alpha Polyclonal specifically detects TNF-alpha in Porcine samples. It is validated for Western Blot, Neutralization, Immunocytochemistry.
Specifications
| TNF-alpha | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 7124 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Neutralization, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Neutralization 0.2-0.8 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| P23563 | |
| TNF | |
| E. coli-derived recombinant porcine TNF-alpha Arg78-Leu232 Accession # P23563 | |
| 25 μg | |
| Primary | |
| Detects porcine TNF-alpha in direct ELISAs and Western blots. | |
| Pig | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction