missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Porcine TNF-alpha Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 4 publications
Brand: R&D Systems AF690
This item is not returnable.
View return policy
Description
TNF-alpha Polyclonal specifically detects TNF-alpha in Porcine samples. It is validated for Western Blot, Neutralization, Immunocytochemistry.
Specifications
| TNF-alpha | |
| Polyclonal | |
| Unconjugated | |
| P23563 | |
| TNF | |
| E. coli-derived recombinant porcine TNF-alpha Arg78-Leu232 Accession # P23563 | |
| 100 μg | |
| Primary | |
| Detects porcine TNF-alpha in direct ELISAs and Western blots. | |
| Pig | |
| IgG |
| Western Blot, Neutralization, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Neutralization 0.2-0.8 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 7124 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction