missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PORCN Polyclonal Antibody

Description
Immunogen sequence: PLNGDRLLRN KKRKARWLRA YESAVSFHFS NYFVGFLSEA TATLAGAGFT EEKDHLEWDL TVSKPLNVEL PRSMVEVVTS WNLPMSYWLN NYVFKNALRL G; Positive Samples: Mouse kidney, Mouse liver, Rat kidney; Cellular Location: Endoplasmic reticulum membrane, Multi-pass membrane protein.
PORCN belongs to the evolutionarily conserved porcupine (Porc) gene family. Genes of the Porc family encode endoplasmic reticulum proteins with multiple transmembrane domains. Porcupine proteins are involved in the processing of Wnt (wingless and int homologue) proteins. Multiple alternatively spliced transcript variants encoding distinct isoforms have been observed.This gene belongs to the evolutionarily conserved porcupine (Porc) gene family. Genes of the Porc family encode endoplasmic reticulum proteins with multiple transmembrane domains. Porcupine proteins are involved in the processing of Wnt (wingless and int homologue) proteins. Multiple alternatively spliced transcript variants encoding distinct isoforms have been observed.
Specifications
Specifications
| Antigen | PORCN |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.46 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | PORCN |
| Gene Accession No. | Q9JJJ7 |
| Gene Alias | 2410004O13Rik; AW045557; DHOF; DXHXS7465e; FODH; Mg61; MGC29687; mMg61; Mporc; por; PORC; Porcn; porcupine; porcupine homolog; porcupine homolog (Drosophila); porcupine O-acyltransferase; Ppn; probable protein-cysteine N-palmitoyltransferase porcupine; Protein MG61; protein-cysteine N-palmitoyltransferase porcupine; protein-serine O-palmitoleoyltransferase porcupine; RGD1564947 |
| Gene Symbols | PORCN |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?