Learn More
Porimin Antibody (CL14381) [PE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26852PE
Product Code. 30296618
  • € 791.00 / 0.10mL
Description
Porimin Monoclonal antibody specifically detects Porimin in Human samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Porimin
Monoclonal
PE
KCT-3, KCT3keratinocytes associated transmembrane protein 3, Keratinocytes-associated transmembrane protein 3, PORIMINporimin, pro oncosis receptor inducing membrane injury, Pro-oncosis receptor inducing membrane injury, serine/threonine-rich receptor, transmembrane protein 123PORMIN
This antibody was developed using a synthetic peptide derived from Q8N131, with the exact immunogen sequence remaining proprietary.
0.1 mL
Apoptosis
114908
Store at 4⪚C in the dark.
IgG2b
Immunohistochemistry
CL14381
PBS
Mouse
Protein A purified