Learn More
Thermo Scientific™ POROS™ GoPure™ 50 HS Pre-packed Column, 0.8 x 10 cm, 5 mL
Tienda Thermo Scientific ProductosDescripción
POROS GoPure 0.8 cm pre-packed columns are intended for the screening of POROS affinity, ion exchange, and hydrophobic interaction chromatography (HIC) resins or for small scale purification.
These pre-packed columns are ready to connect and have the following features:
- Dimensions of 0.8 x 10 cm, with a nominal bed volume of 5 mL, providing a uniform screening platform
- Direct connection to standard chromatography systems
- Convenient—ready-to-run columns simplify and speed POROS resin evaluations
- The 10-cm bed height and 5-mL bed volume allow initial process development at a bench scale
- Packed with POROS high performance resins that offer high dynamic binding capacity and high resolution over a wide range of flow rates
The POROS HS 50 μm resin (Cat. No. 1335906) in this POROS GoPure column has a sulphopropyl functional group for strong cation exchange chromatography.
Other POROS anion exchange resins, as well cation exchange resins, are also available in this column format. All POROS High Performance Chromatography resins have a unique pore structure that provides excellent mass transport for high capacity and resolution over a wide range of flow rates.
Especificaciones
Especificaciones
| Column Capacity | 5 mL |
| Length (Metric) | 10 cm |
| Quantity | 1 column |
| Type | Prepacked Column |
| Diameter (Metric) Inner | 0.8 cm |
| Product Line | POROS GoPure |
| Shipping Condition | Room Temperature |
| Column Format | Pre-packed |