Learn More
Prestige Medical™ Portable Autoclave Classic Media
Product Code. 15717572
Change view
Click to view available options
Capacity (Metric):
12 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
15717572 12 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15717572 Supplier Prestige Medical™ Supplier No. 210048
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use this autoclave where a cost effective, portable autoclave is needed. Prestige Medical™ Portable Autoclave Classic Media is fully automatic, simply press the button to start. Lightweight and portable, ideal for laboratory use. This 12 liter Classic Media autoclave sterilizes at 121°C.
  • High quality construction
  • Fully automatic electronic time and temperature control
  • Steam release valve automatically opens to release excess pressure
  • Automatic fault warning light and cut-out if unit boils dry or sterilisation is interrupted
  • Dual interlock handles prevent the lid being opened before the pressure and temperature inside have fallen to safe levels
  • Complete with instrument basket
  • Models 2015 onwards available with optional printer for easy data recording
  • CE marked
  • Comprehensive one year warranty (UK)
  • Interlock safety mechanism, eliminates the risk of super-heated vessels exploding
  • Reliable, special in-built software ensures exact temperatures are reached for the correct duration
  • 9L and 12L models
  • Fits neatly onto the smallest of workbenches For the sterilization of media, liquids and contaminated waste
Specifications
Capacity (Metric) 12 L