missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POT1 Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB500-176JF549
This item is not returnable.
View return policy
Description
POT1 Polyclonal antibody specifically detects POT1 in Human samples. It is validated for Western Blot
Specifications
| POT1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Janelia Fluor 549 | |
| DKFZp586D211, hPot1, POT1 protection of telomeres 1 homolog, POT1-like telomere end-binding protein, protection of telomeres 1 homolog (S. pombe), protection of telomeres protein 1 | |
| A synthetic peptide made to an internal portion of the human POT1 protein sequence (between residues 200-300). [UniProt# Q9NUX5] | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Chromatin Research, DNA replication Transcription Translation and Splicing | |
| 25913 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction