Learn More
Potassium 2-methylthiophenyltrifluoroborate, 96%, Thermo Scientific™
Product Code. 15449347
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15449347 1 g 1g
15459347 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15449347 Supplier Thermo Scientific Alfa Aesar Supplier No. H52587.03
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 850623-77-7
Molecular Formula C7H7BF3KS
Molecular Weight (g/mol) 230.097
MDL Number MFCD04115773
InChI Key ACOYJOWMHXJLJP-UHFFFAOYSA-N
Synonym potassium trifluoro 2-methylthio phenyl borate, potassium 2-methylthiophenyl trifluoroborate, potassium trifluoro 2-methylsulfanyl phenyl boranuide, potassium 2-methylsulfanylphenyl trifluoroborate, potassium 2-methylthiophenyltrifluoroborate, potassium trifluoro-2-methylthio phenyl boranuide, potassium trifluoro 2-methylsulfanyl phenyl borate 1-, potassium ion trifluoro 2-methylsulfanyl phenyl boranuide, potassium tris fluoranyl-2-methylsulfanylphenyl boranuide
PubChem CID 44717213
IUPAC Name potassium;trifluoro-(2-methylsulfanylphenyl)boranuide
SMILES [B-](C1=CC=CC=C1SC)(F)(F)F.[K+]
Specifications
Melting Point 274°C to 276°C
Quantity 1 g
Formula Weight 230.1
Percent Purity 96%
Chemical Name or Material Potassium 2-methylthiophenyltrifluoroborate