Learn More
Potassium 4-fluorophenyltrifluoroborate, 95%
Product Code. 11378596
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11378596 5 g 5g
11368596 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11378596 Supplier Thermo Scientific Alfa Aesar Supplier No. L17655.06
 Restricted Item
Estimated Shipment: 26-08-2026
Add to basket
CAS: 192863-35-7 | C6H4BF4K | 202 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 192863-35-7
Molecular Formula C6H4BF4K
Molecular Weight (g/mol) 202
MDL Number MFCD01318170
InChI Key PXKNUQDMYBUYSZ-UHFFFAOYSA-N
Synonym potassium 4-fluorophenyltrifluoroborate, potassium trifluoro 4-fluorophenyl borate, potassium trifluoro 4-fluorophenyl boranuide, potassium 4-fluorophenyl trifluoroborate, 4-fluorophenyl-trifluoroborate potassium salt, 4-fluorophenyltrifluoroboric acid potassium salt, 4-fluorobenzenetrifluoroboric acid potassium salt, pubchem11422, potassiotrifluoro 4-fluorophenyl boron v, potassium trifluoro-4-fluorophenyl boranuide
PubChem CID 23677644
IUPAC Name potassium;trifluoro-(4-fluorophenyl)boranuide
SMILES [B-](C1=CC=C(C=C1)F)(F)(F)F.[K+]
Specifications
Melting Point 290°C to 292°C
Quantity 5 g
Beilstein 7783544
Formula Weight 202
Percent Purity 95%
Chemical Name or Material Potassium 4-fluorophenyltrifluoroborate