Learn More
Potassium 4-n-butoxyphenyltrifluoroborate, 96%, Thermo Scientific™
Product Code. 15459867
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15459867 1 g 1g
15440366 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15459867 Supplier Thermo Scientific Alfa Aesar Supplier No. H53092.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 850623-61-9
Molecular Formula C10H13BF3KO
Molecular Weight (g/mol) 256.117
MDL Number MFCD04115755
InChI Key UEVIEKRZQJRPGE-UHFFFAOYSA-N
Synonym potassium 4-butoxyphenyl trifluoroborate, potassium 4-butoxyphenyltrifluorate, potassium 4-butoxyphenyl trifluoroboranuide, potassium 4-n-butoxyphenyl trifluorate, potassium 4-n-butoxyphenyl trifluoroborate, potassium 4-butoxyphenyl-trifluoroboranuide, potassium 4-butoxyphenyl trifluoro borate 1-, potassium 4-n-butoxyphenyltrifluoroborate, potassium ion 4-butoxyphenyl trifluoroboranuide, potassium 4-butoxyphenyl-tris fluoranyl boranuide
PubChem CID 44717219
IUPAC Name potassium;(4-butoxyphenyl)-trifluoroboranuide
SMILES [B-](C1=CC=C(C=C1)OCCCC)(F)(F)F.[K+]
Specifications
Melting Point >300°C
Quantity 1 g
Formula Weight 256.12
Percent Purity 96%
Chemical Name or Material Potassium 4-n-butoxyphenyltrifluoroborate