missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium chloride, 0.159M, Conductivity Standard
Click to view available options
Quantity:
500 mL
1 L
4 L
Unit Size:
1 L
4 L
500mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Vapor Pressure | 23 hPa (17mm Hg) at 20°C |
| Density | 1.01181g/cm3 at 20°C |
| Assay Percent Range | 0.159M |
| Physical Form | Liquid |
| Color | Colorless |
| IUPAC Name | potassium chloride |
| Quantity | 1 L |
| Formula Weight | 74.55 |
| Concentration or Composition (by Analyte or Components) | Potassium chloride: 1.2%; Water: 98.8% |
| Chemical Name or Material | Potassium chloride, 20, 000μmho at 25°C |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction