missing translation for 'onlineSavingsMsg'
Learn More

Potassium Cyanide-13C, TRC

Product Code. 30413681
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30413681 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30413681 Supplier Toronto Research Chemicals Supplier No. TRCP6950521G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Potassium Cyanide-13C
CAS 25909-68-6
Molecular Formula 13C K N
Quantity 1 g
Synonym Hydrocyanic-13C acid','Potassium salt, Potassium [13C]cyanide, Potassium Cyanide-13C
InChI Formula InChI=1S/CN.K/c1-2;/i1+1;
SMILES [K][13C]#N
IUPAC Name azanylidyne(1^{13}C)methylpotassium
Molecular Weight (g/mol) 66.11
Formula Weight 65.9701
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.